Learn More
Eppendorf™ Adapters For Fixed-Angle Round Bottom Rotor
Shop All Eppendorf Products
Click to view available options
Capacity:
15 to 18 mL
5 mL
7 to 15 mL
Unit Size:
Pack of 2
Description
- Adapters
- For Eppendorf™ Fixed Angle Round-Bottom Rotors
Specifications
Specifications
Type | Adapter |
Tube Type | Round Bottom |
Capacity | 15 to 18 mL |
For Use With (Equipment) | Fixed angle rotor |
Bore Size (Metric) | 18 mm |
No. of Places | 6 |
No. per Pack | 2 Pack |
Product Content Correction
Product Title
Eppendorf™ Adapters For Fixed-Angle Round Bottom Rotor >