Learn More
Hettich™ Adapter for Carriers no. 4295-A and 4522-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
1390.00 NOK - 2790.00 NOK
Specifications
Type Adapter
For Use With (Equipment) Carriers no. 4295-A and 4522-A
View More Specs
Products 8
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
12043904
View Documents Promotion Details
Hettich™
4222
4 to 7 mL Tubes
1450.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
10247762
View Documents Promotion Details
Hettich™
4214-93
30 x 2.7 to 5 mL Tubes
1390.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
10788315
View Documents Promotion Details
Hettich™
4213-93
2.7 to 5 mL Tubes
1390.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
12073904
View Documents Promotion Details
Hettich™
4224
63 x 4 mL
1695.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
11445018
View Documents Promotion Details
Hettich™
4238
250 to 290 mL
2040.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
12083904
View Documents Promotion Details
Hettich™
4258
500 to 750 mL
2310.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
11465018
View Documents Promotion Details
Hettich™
4249
6 x 50 mL
2090.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
11485178
View Documents Promotion Details
Hettich™
4245-A
8 x 50 mL
2790.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
Description
  • Centrifuge Adapters for ROTO SILENTA 630 RS and ROTIXA 500 RS Centrifuges
Specifications
Specifications
Adapter
Carriers no. 4295-A and 4522-A