Learn More
Hettich™ Adapter for Carriers no. 4295-A and 4522-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4214-93
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10247762
  • 1390.00 NOK / Each
Estimated Shipment: 19-06-2024
Description
  • Centrifuge Adapters for ROTO SILENTA 630 RS and ROTIXA 500 RS Centrifuges
Specifications
Specifications
Adapter
30 x 2.7 to 5 mL Tubes
75 mm
30
Blood/Urine Tubes
16 mm
Carriers no. 4295-A and 4522-A