Learn More
Hettich™ Adapter for Carriers no. 4295-A and 4522-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 4214-93
Additional Details : Weight : 0.01000kg
Description
- Centrifuge Adapters for ROTO SILENTA 630 RS and ROTIXA 500 RS Centrifuges
Specifications
Adapter | |
30 x 2.7 to 5 mL Tubes | |
75 mm | |
30 |
Blood/Urine Tubes | |
16 mm | |
Carriers no. 4295-A and 4522-A |