missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Adapter for Carriers no. 4295-A and 4522-A

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 4258

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12083904

  • 2310.00 NOK / Each
Estimated Shipment: 09-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Centrifuge Adapters for ROTO SILENTA 630 RS and ROTIXA 500 RS Centrifuges
Specifications

Specifications

Adapter
500 to 750 mL
147 mm
1
Corning Tubes and Plastic Bottles
97 mm
Carriers no. 4295-A and 4522-A
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers