Learn More
Hettich™ Adapter for Carriers no. 4295-A and 4522-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4224
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12073904
  • 1695.00 NOK / Each
Estimated Shipment: 09-07-2024
Description
  • Centrifuge Adapters for ROTO SILENTA 630 RS and ROTIXA 500 RS Centrifuges
Specifications
Specifications
Adapter
63 x 4 mL
88 mm
63
Round Bottom
10 mm
Carriers no. 4295-A and 4522-A