accessibility menu, dialog, popup

UserName

CH3H3CCH2HNBrHNCH3HCl

CAS RN 951322-11-5

IUPAC Name: {2-[6-bromo-2-(2-methylbut-3-en-2-yl)-1H-indol-3-yl]ethyl}(methyl)amine hydrochloride
Synonyms: desformylflustrabromine
Molecular Weight (g/mol): 357.72
Molecular Formula: C16H22BrClN2
InChi Key: GEZWEAPBLKXKGN-UHFFFAOYSA-N
SMILES: Cl.CNCCC1=C(NC2=CC(Br)=CC=C12)C(C)(C)C=C
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results