Learn More
Desformylflustrabromine hydrochloride, MedChemExpress
Product Code. 16495106
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495106 100 mg 100mg
16482966 25 mg 25mg
16440497 50 mg 50mg
16434148 10 mg 10mg
16416857 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16495106 Supplier Medchem Express Supplier No. HY107675
MedChemExpress Desformylflustrabromine hydrochloride is a selective agonist of α4β2 neuronal nicotinic acetylcholine receptor (nAChR) with a pEC50 of 6.48.
Desformylflustrabromine hydrochloride is a selective agonist of α4β2 neuronal nicotinic acetylcholine receptor (nAChR) with a pEC50 of 6.48.
Specifications
Chemical Name or Material Desformylflustrabromine hydrochloride
CAS 951322-11-5
Color Pink
Purity Grade Notes Research
Molecular Formula C16H22BrClN2
Quantity 100 mg
Synonym Deformylflustrabromine hydrochloride dFBr hydrochloride
Solubility Information DMSO : ≥ 105 mg/mL (293.53 mM) ∣H2O : 5 mg/mL (13.98 mM; Need ultrasonic)
SMILES CNCCC1=C(C(C)(C)C=C)NC2=CC(Br)=CC=C12.[H]Cl
Molecular Weight (g/mol) 357.72
Formula Weight 357.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation