accessibility menu, dialog, popup

UserName

CAS RN 916170-19-9

CH3NNHONNHONHOBr(E/Z)

CAS RN 916170-19-9

IUPAC Name: N-({N'-[1-(3-bromophenyl)ethylidene]hydrazinecarbonyl}(4-oxo-3,4-dihydrophthalazin-1-yl)methyl)benzamide
Synonyms: N-({N'-[1-(3-bromophenyl)ethylidene]hydrazinecarbonyl}(4-oxo-3H-phthalazin-1-yl)methyl)benzamide
Molecular Weight (g/mol): 518.37
Molecular Formula: C25H20BrN5O3
InChi Key: UCUHFWIFSHROPY-UHFFFAOYNA-N
SMILES: CC(=NNC(=O)C(NC(=O)C1=CC=CC=C1)C1=NNC(=O)C2=CC=CC=C12)C1=CC(Br)=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results