Learn More
AC-55541, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
AC-55541 is a highly selective protease-activated receptor 2 (PAR2) agonist (pEC50=6.7), displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. AC-55541 has pEC50 values of 5.9 and 6.6 in PI hydrolysis assays and Ca2+ mobilization assays and exhibits pronociceptive activity in vivo.
Specifications
Specifications
| Chemical Name or Material | AC-55541 |
| CAS | 916170-19-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H20BrN5O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 51 mg/mL (98.39 mM) |
| SMILES | O=C(NC(C(C1=CC=CC=C12)=NNC2=O)C(N/N=C(C)/C3=CC=CC(Br)=C3)=O)C4=CC=CC=C4 |
| Molecular Weight (g/mol) | 518.36 |
| Formula Weight | 518.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.