CAS RN 91396-88-2
CAS RN 91396-88-2
IUPAC Name:
N'-phenylpyridine-4-carbohydrazide
Synonyms:
N'-phenylpyridine-4-carbohydrazide
Molecular Weight (g/mol):
213.24
Molecular Formula:
C12H11N3O
InChi Key:
HUDWXDLBWRHCKO-UHFFFAOYSA-N
SMILES:
O=C(NNC1=CC=CC=C1)C1=CC=NC=C1
1
–
2
of
2
results
1
PluriSln 1, Tocris Bioscience™
CAS: 91396-88-2 Molecular Formula: C12H11N3O Molecular Weight (g/mol): 213.24 MDL Number: MFCD02344904 InChI Key: HUDWXDLBWRHCKO-UHFFFAOYSA-N Synonym: plurisln 1,n'-phenylisonicotinohydrazide,plurisln 1 nsc 14613,plurisin 1,plurisin #1 nsc 14613,plurisin #1,4-pyridinecarboxylicacid, 2-phenylhydrazide,4-pyridinecarboxylic acid 2-phenylhydrazide,plurisln,plurisln1 PubChem CID: 225362 IUPAC Name: N'-phenylpyridine-4-carbohydrazide SMILES: O=C(NNC1=CC=CC=C1)C1=CC=NC=C1
2
PluriSIn 1, MedChemExpress
MedChemExpress PluriSIn 1 (NSC 14613) is an inhibitor of stearoyl-coA desaturase (SCD), and is a pluripotent cell-specific inhibitor.
