accessibility menu, dialog, popup

UserName

ClNNNNCl

CAS RN 74115-24-5

IUPAC Name: bis(2-chlorophenyl)-1,2,4,5-tetrazine
Synonyms: clofentezine caswell no. 593A acaristop apollo apollo (pesticide) apollo 50W bisclofentazin panatac bisclofentazine epa pesticide chemical code 125501
Molecular Weight (g/mol): 303.15
Molecular Formula: C14H8Cl2N4
InChi Key: UXADOQPNKNTIHB-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C1=NN=C(N=N1)C1=CC=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Concentration 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results