Learn More
Clofentezine in Methanol 100μg/mL, Fisher Chemical™
BIOPHARMA
Product Code. 12953234 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12953234 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12953234 Supplier Fisher Chemical Supplier No. P874910
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 74115-24-5
Molecular Formula C14H8Cl2N4
Concentration 100 μg/mL
InChI Key UXADOQPNKNTIHB-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C2=NN=C(N=N2)C3=CC=CC=C3Cl)Cl
IUPAC Name 3,6-bis(2-chlorophenyl)-1,2,4,5-tetrazine
Solvent or Matrix Methanol
Molecular Weight (g/mol) 303.146
PubChem CID 73670
Quantity 1 mL
ChEBI CHEBI:39315
Chemical Name or Material Clofentezine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Metabolite Clofentezine in Methanol 100µg/mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol