Learn More
Clofentezine, MedChemExpress
Description
Clofentezine, a growth inhibitor, has sublethal effects on life-table parameters of Tetranychus urticae Koch females.
Specifications
Specifications
| Chemical Name or Material | Clofentezine |
| Physical Form | Solid |
| CAS | 74115-24-5 |
| Color | Pale Purple |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H8Cl2N4 |
| Solubility Information | DMSO : 15.5 mg/mL (51.13 mM; Need ultrasonic and warming) |
| SMILES | ClC1=CC=CC=C1C2=NN=C(C3=CC=CC=C3Cl)N=N2 |
| Molecular Weight (g/mol) | 303.15 |
| Show More |