Learn More
Clofentezine, TRC
Product Code. 30362954
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362954 1 g 1g
30352669 10 g 10g
30355706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30362954 Supplier Toronto Research Chemicals Supplier No. TRCC5856151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74115-24-5
Molecular Formula C14 H8 Cl2 N4
Molecular Weight (g/mol) 303.15
Synonym 3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine, 3,6-Di-2-chlorophenyl-1,2,4,5-tetrazine, Acaristop, Apollo, Apollo (pesticide), Apollo 50W, Apollo SOSC, Bisclofentezin, Bisclofentezine, Clofentezine, ManSiJing, NC 21314
IUPAC Name 3,6-bis(2-chlorophenyl)-1,2,4,5-tetrazine
SMILES Clc1ccccc1c2nnc(nn2)c3ccccc3Cl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H8Cl2N4/c15-11-7-3-1-5-9(11)13-17-19-14(20-18-13)10-6-2-4-8-12(10)16/h1-8H
Formula Weight 302.0126
Chemical Name or Material Clofentezine