accessibility menu, dialog, popup

UserName

CAS RN 724741-75-7

CH3H3CCH3SOOHNONHN

CAS RN 724741-75-7

IUPAC Name: 4-[(4-tert-butylbenzenesulfonamido)methyl]-N-(pyridin-3-yl)benzamide
Synonyms: 4-[(4-tert-butylbenzenesulfonamido)methyl]-N-(pyridin-3-yl)benzamide
Molecular Weight (g/mol): 423.53
Molecular Formula: C23H25N3O3S
InChi Key: NGQPRVWTFNBUHA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)NCC1=CC=C(C=C1)C(=O)NC1=CN=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results