CAS RN 645-66-9
CAS RN 645-66-9
IUPAC Name:
dodecanoyl dodecanoate
Synonyms:
dodecanoic acid, anhydride dodecenoyl anhydride
Molecular Weight (g/mol):
382.63
Molecular Formula:
C24H46O3
InChi Key:
NWADXBLMWHFGGU-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC
1
–
2
of
2
results
2
Lauric anhydride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.