Learn More
Lauric Anhydride, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
5g
Specifications
Specifications
| CAS | 645-66-9 |
| Molecular Formula | C24H46O3 |
| Quantity | 10 g |
| Synonym | lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride |
| InChI Key | NWADXBLMWHFGGU-UHFFFAOYSA-N |
| SMILES | CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC |
| IUPAC Name | dodecanoyl dodecanoate |
| Molecular Weight (g/mol) | 382.629 |
| PubChem CID | 69527 |
| Chemical Name or Material | Lauric Anhydride |