Learn More
Lauric anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | Lauric anhydride |
| CAS | 645-66-9 |
| Molecular Formula | C24H46O3 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C24H46O3/c1-3-5-7-9-11-13-15-17-19-21-23(25)27-24(26)22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3 |
| SMILES | CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC |
| IUPAC Name | dodecanoyl dodecanoate |
| Molecular Weight (g/mol) | 382.62 |
| Formula Weight | 382.3447 |