CAS RN 614749-78-9
CAS RN 614749-78-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ SM 16
Potent TGF-β receptor type 1 (TGF-βRI) inhibitor
2
SM 16, MedChemExpress
MedChemExpress SM 16 is a ALK5/ALK4 kinase inhibitor with Kis of 10 and 1.5 nM, respectively.