Learn More
SM 16, MedChemExpress
Product Code. 16467187
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467187 5 mg 5mg
16460777 50 mg 50mg
16435346 100 mg 100mg
16424468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16467187 Supplier Medchem Express Supplier No. HY111482
MedChemExpress SM 16 is a ALK5/ALK4 kinase inhibitor with Kis of 10 and 1.5 nM, respectively.
SM 16 is a ALK5/ALK4 kinase inhibitor with Kis of 10 and 1.5 nM, respectively.
Specifications
Chemical Name or Material SM 16
CAS 614749-78-9
Color White
Purity Grade Notes Research
Molecular Formula C25H26N4O3
Quantity 5 mg
Solubility Information DMSO : 65 mg/mL (150.99 mM; Need ultrasonic)
SMILES O=C(C1(CC2)CCC2(C3=NC(C4=CC=C(OCO5)C5=C4)=C(C6=NC(C)=CC=C6)N3)CC1)N
Molecular Weight (g/mol) 430.5
Formula Weight 430.5
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease