accessibility menu, dialog, popup

UserName

CAS RN 3366-95-8

CH3OHNCH3NNOO

CAS RN 3366-95-8

IUPAC Name: 1-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol
Synonyms: secnidazole
Molecular Weight (g/mol): 185.18
Molecular Formula: C7H11N3O3
InChi Key: KPQZUUQMTUIKBP-UHFFFAOYNA-N
SMILES: CC(O)CN1C(C)=NC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
  • (2)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)