Learn More
Secnidazole, LoGiCal
Product Code. 30459935
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459935 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459935 Supplier LoGiCal Supplier No. LGCFOR0119.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Secnidazole
CAS 3366-95-8
Analyte or Component Names Secnidazole
Shipping Condition Room Temperature
Molecular Formula C7 H11 N3 O3
Certifications/Compliance ISO 9001
Synonym Secnidazole
InChI Formula InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3
Solution Type Neat
SMILES CC(O)Cn1c(C)ncc1[N+](=O)[O-]
IUPAC Name 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
Industry Type Clinical Testing
Molecular Weight (g/mol) 185.18
Quantity 10 mg
Formula Weight 185.08 g/mol
Show More Show Less
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.