Learn More
Secnidazole, Mikromol™
Product Code. 17911766
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17911766 Supplier Mikromol Supplier No. MM0119.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Secnidazole
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Secnidazole
CAS 3366-95-8
Analyte or Component Names Secnidazole
Shipping Condition Room Temperature
Molecular Formula C7 H11 N3 O3
Certifications/Compliance ISO 17034
Synonym Secnidazole
InChI Formula InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3
SMILES CC(O)Cn1c(C)ncc1[N+](=O)[O-]
IUPAC Name 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
Molecular Weight (g/mol) 185.18
Quantity 100 mg
Formula Weight 185.08 g/mol