Learn More
Secnidazole 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30323388
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323388 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323388 Supplier Dr. Ehrenstorfer Supplier No. DREA16931100AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Secnidazole
CAS 3366-95-8
Analyte or Component Names Secnidazole
Shipping Condition Room Temperature
Molecular Formula C7 H11 N3 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Secnidazole
InChI Formula InChI=1S/C7H11N3O3/c1-5(11)4-9-6(2)8-3-7(9)10(12)13/h3,5,11H,4H2,1-2H3
SMILES CC(O)Cn1c(C)ncc1[N+](=O)[O-]
IUPAC Name 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 185.18
Quantity 1 mL
Formula Weight 185.08 g/mol