accessibility menu, dialog, popup

UserName

CAS RN 290315-45-6

CH3FOOHNFFSOOCl

CAS RN 290315-45-6

IUPAC Name: 4-(2-{1-[N-(2,5-difluorophenyl)4-chlorobenzenesulfonamido]ethyl}-5-fluorophenyl)butanoic acid
Synonyms: 4-(2-{1-[N-(2,5-difluorophenyl)4-chlorobenzenesulfonamido]ethyl}-5-fluorophenyl)butanoic acid
Molecular Weight (g/mol): 511.94
Molecular Formula: C24H21ClF3NO4S
InChi Key: IZAOBRWCUGOKNH-UHFFFAOYNA-N
SMILES: CC(N(C1=C(F)C=CC(F)=C1)S(=O)(=O)C1=CC=C(Cl)C=C1)C1=C(CCCC(O)=O)C=C(F)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results