missing translation for 'onlineSavingsMsg'
Learn More

BMS 299897, MedChemExpress

Product Code. p-200098070
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16488128 10 mg 10mg
16434047 50 mg 50mg
2 options
This item is not returnable. View return policy

Product Code. 16488128

Brand: Medchem Express HY50883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS 299897 is a sulfonamide γ-secretase inhibitor with an IC50 of 7 nM for Aβ production inhibition in HEK293 cells stably overexpressing amyloid precursor protein (APP).

BMS 299897 is a sulfonamide γ-secretase inhibitor with an IC50 of 7 nM for Aβ production inhibition in HEK293 cells stably overexpressing amyloid precursor protein (APP).

Specifications

Chemical Name or Material BMS 299897
CAS 290315-45-6
Color White
Purity Grade Notes Research
Molecular Formula C24H21ClF3NO4S
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (58.60 mM)
SMILES FC1=CC=C(F)C(N([C@@H](C2=C(CCCC(O)=O)C=C(F)C=C2)C)S(C3=CC=C(Cl)C=C3)(=O)=O)=C1
Molecular Weight (g/mol) 511.94
Formula Weight 511.94
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.