accessibility menu, dialog, popup

UserName

CAS RN 212844-53-6

CH3CH3(R)HONHNNHClNNCH3H3CN

CAS RN 212844-53-6

IUPAC Name: (2R)-2-({6-[(3-chlorophenyl)amino]-9-(propan-2-yl)-9H-purin-2-yl}amino)-3-methylbutan-1-ol
Synonyms: purvalanol A C19H25clN6O Purv
Molecular Weight (g/mol): 388.9
Molecular Formula: C19H25ClN6O
InChi Key: PMXCMJLOPOFPBT-HNNXBMFYSA-N
SMILES: CC(C)[C@H](CO)NC1=NC(NC2=CC=CC(Cl)=C2)=C2N=CN(C(C)C)C2=N1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results