accessibility menu, dialog, popup

UserName

CAS RN 1622849-58-4

H3CNOONHONO

CAS RN 1622849-58-4

IUPAC Name: 5-benzyl-N-(5-methyl-4-oxo-2,3,4,5-tetrahydro-1,5-benzoxazepin-3-yl)-1,2-oxazole-3-carboxamide
Synonyms: 5-benzyl-N-(5-methyl-4-oxo-2,3-dihydro-1,5-benzoxazepin-3-yl)-1,2-oxazole-3-carboxamide
Molecular Weight (g/mol): 377.4
Molecular Formula: C21H19N3O4
InChi Key: KNOUWGGQMADIBV-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2OCC(NC(=O)C2=NOC(CC3=CC=CC=C3)=C2)C1=O
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results