Learn More
Thermo Scientific™ GSK481, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | GSK481 |
| CAS | 1622849-58-4 |
| Synonym | 5-(Phenylmethyl)-N-[(3 S)-2,3,4,5-tetrahydro-5-methyl-4-oxo-1,5-benzoxazepin-3-yl]-3-isoxazolecarboxamide |
| Quantity | 100 mg |
| Molecular Formula | C21 H19 N3 O4 |
| Formula Weight | 377.40 |
| Physical Form | Solid |
| Product Type | Reagent |
| Purity | 95% |