Learn More
Tocris Bioscience™ XCC
GREENER_CHOICE
Product Code. 18035602
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18035602 10 mg 10mg
18064222 50 mg 50mg
2 options
Product Code. 18035602
Brand: Tocris Bioscience™ 4220/10
Adenosine receptor antagonist
Adenosine receptor antagonist (Ki values are 42, 68 and 1130 nM for A1, A2B and A2A receptors).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 96865-83-7
Molecular Formula C19H22N4O5
Specifications
Quantity 10 mg
Formula Weight 386.4
Purity >98%
Molecular Weight (g/mol) 386.408
Chemical Name or Material XCC