missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WZB 117

Description
WZB 117 is a GLUT inhibitor (Ki(app) values are 0.2, 10 and 10 μM for GLUT4, GLUT1 and GLUT3, respectively). Competitive inhibitor of net glucose uptake but a noncompetitive inhibitor of sugar efflux from cells. Inhibits passive sugar transport in human erythrocytes and cancer cell lines. Attenuates erythrocyte 3-O-metylglucose (3MG) uptake in vitro (Ki = 6 μM). Suppresses tumor growth in a nude mouse model.
Specifications
Specifications
| Chemical Name or Material | 3-Fluoro-1,2-phenylene bis(3-hydroxybenzoate) |
| CAS | 1223397-11-2 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C20H13FO6 |
| Formula Weight | Observed MW: 368.31 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?