Learn More
Fisherbrand™ Wire Mesh Basket
Product Code. p-7221109 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
100 x 100 x 100 mm
120 x 120 x 120 mm
140 x 140 x 140 mm
150 x 150 x 150 mm
160 x 160 x 160 mm
180 x 180 x 180 mm
200 x 200 x 150 mm
200 x 200 x 200 mm
250 x 150 x 150 mm
250 x 250 x 250 mm
300 x 200 x 150 mm
300 x 200 x 200 mm
300 x 300 x 300 mm
400 x 200 x 150 mm
400 x 200 x 200 mm
Unit Size:
Each
Product Code. 17187914
Only null left
Add to basket
Add to basket
Basket that is useful for small instruments.
  • For small instruments
  • Available in different sizes
Specifications
Mesh Size 8 x 8 x 1 mm
For Use With (Equipment) Small Instruments
Dimensions (L x W x H) 100 x 100 x 100 mm