missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WIC1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
WIC1 is an inhibitor of the canonical Wnt/β-catenin signaling pathway. In vitro, it reduces ABSC (Airway Basal Stem Cell) proliferation and induces ciliated epithelial cell differentiation, in a model of squamous lung cancer. WIC1 also represses the phosphorylation of β-cateninY489 and Tp63 expression.
Specifications
Specifications
| Chemical Name or Material | N-[4-(4-Ethyl-1-piperazinyl)phenyl]-2-oxo-2 H-1-benzopyran-3-carboxamide |
| CAS | 943083-58-7 |
| Quantity | 10 mg |
| Target | beta-Catenin |
| Molecular Formula | C22H23N3O3 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction