Learn More
DWK Life Sciences Wheaton™ 25mL Erlenmeyer Flask With 20-400 Screw Threads
Brand:  DWK Life Sciences 635134
270.33 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12467121
  • 385.00 NOK / Each
Only null left
Estimated Shipment: 15-01-2026
Specifications
25 mL
Borosilicate Glass
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ 25mL Erlenmeyer Flask With 20-400 Screw Threads > 25mL