Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
Brand:  DWK Life Sciences 356902
1926.99 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
2576.94 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12441421
  • 3025.00 NOK / Each
Only null left
Estimated Shipment: 12-09-2025
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500mL Celstir Spinner Flask
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Impeller
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks > Impeller, For Flask: 500mL