Learn More
DWK Life Sciences Wheaton™ E-Z Decappers
Product Code. p-4879904 Shop All Wheaton Science Products Products
11 mm Seals, Each
Click to view available options
For Use With (Equipment):
11 mm Seals
13 mm Seals
20 mm Seals
8 mm Seals
Unit Size:
Each
Product Code. 10785183
Brand: DWK Life Sciences W225351
Only undefined left
Only undefined left
Add to basket
Add to basket
With ergonomically designed handles for sealing and decapping standard laboratory vials
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
  • Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
  • The decappers come with visible size labels for easy identification.
  • User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
  • Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Warranty
Two years
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 11 mm Seals
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ E-Z Decappers >