Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
These closures for ABC Vials feature the Wheaton™ Super Septa, which is low extractable and optimal for GC/MS.
4350.00 NOK - 8025.00 NOK
Product Code | Brand | Color | Pre-slit | Septum | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Pre-slit | Septum | Price | Quantity & Availability | |||
12425051
|
DWK Life Sciences™
W225338-08 |
Natural | Yes | PTFE, Silicone |
8025.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12425041
|
DWK Life Sciences™
W225332-0203 |
Red | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12405041
|
DWK Life Sciences™
W225332-02 |
Natural | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12435041
|
DWK Life Sciences™
W225332-0204 |
Blue | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12415051
|
DWK Life Sciences™
W225336-0601 |
Black | No | PTFE |
4350.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12465041
|
DWK Life Sciences™
W225333-03 |
Natural | No | PTFE, Silicone, PTFE |
7030.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12415041
|
DWK Life Sciences™
W225332-0201 |
Black | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12445041
|
DWK Life Sciences™
W225332-0205 |
Yellow | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
12455041
|
DWK Life Sciences™
W225332-0207 |
Green | No | PTFE, Silicone |
5270.00 NOK
Pack of 1000 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||
Description
- Screw caps are available in natural, black, red, blue, yellow or green polypropylene
Specifications
Polypropylene | |
100 Caps/Pk., 10 Pk./Cs. |