Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
Product Code. 12465041 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
Septum:
PTFE
PTFE, Silicone
PTFE, Silicone, PTFE
Pre-slit:
No
Yes
Unit Size:
Pack of 1000
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Septum Pre-slit unitSize
12465041 Natural PTFE, Silicone, PTFE No Pack of 1000
12425051 Natural PTFE, Silicone Yes Pack of 1000
12425041 Red PTFE, Silicone No Pack of 1000
12405041 Natural PTFE, Silicone No Pack of 1000
12435041 Blue PTFE, Silicone No Pack of 1000
12415051 Black PTFE No Pack of 1000
12415041 Black PTFE, Silicone No Pack of 1000
12445041 Yellow PTFE, Silicone No Pack of 1000
12455041 Green PTFE, Silicone No Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12465041 Supplier DWK Life Sciences™ Supplier No. W22533303
Only null left
Add to basket
Add to basket
These closures for ABC Vials feature the Wheaton™ Super Septa, which is low extractable and optimal for GC/MS.
  • Screw caps are available in natural, black, red, blue, yellow or green polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Polypropylene
Pre-slit No
Septum PTFE, Silicone, PTFE
Quantity 100 Caps/Pk., 10 Pk./Cs.
Closure Size 9 mm