Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
Shop All Wheaton Science Products ProductsDescription
- Screw caps are available in natural, black, red, blue, yellow or green polypropylene
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| Pre-slit | No |
| Septum | PTFE, Silicone, PTFE |
| Quantity | 100 Caps/Pk., 10 Pk./Cs. |
| Closure Size | 9 mm |