Learn More
DWK Life Sciences™ Wheaton™ Caps for 12 x 32 mm Large Opening ABC Vials
These closures for ABC Vials feature the Wheaton™ Super Septa, which is low extractable and optimal for GC/MS.
Brand:  DWK Life Sciences™ W225332-0204
Additional Details : Weight : 0.36290kg
 View more versions of this product
Product Code. 12435041
  • 5270.00 NOK / Pack of 1000
Estimated Shipment: 25-09-2024
Description
  • Screw caps are available in natural, black, red, blue, yellow or green polypropylene
Specifications
Specifications
Blue
No
100 Caps/Pk., 10 Pk./Cs.
Polypropylene
PTFE, Silicone
9 mm