Learn More
Cytiva Whatman™ WT 100 Witt's Flask Product Code.: 11360834
Only undefined left
Add to basket
Cytiva Whatman™ WT 100 Witt's Flask
Product Code. 11360834
Witts flask
Click to view available options
:
Witts flask
Product Code. 11360834
Brand: Cytiva 10477601
Only undefined left
Add to basket
Only undefined left
Add to basket
WT 100 Witt's Flask, 1000 ml with tubing nozzle, vacuum filtration apparatus accessories, for filtrate collection in an inserted container.
The bottle is made of borosilicate glass. It has a replaceable round lid and side-mounted tubing nozzle for vacuum tubing 8 mm (inside diameter).
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vacuum Filtration Apparatus
Material Borosilicate Glass
Includes Replaceable round lid, side-mounted tubing nozzle for vacuum tubing 8mm
Type Witt's Flask
Volume (Metric) 1000 mL
Height (Metric) 160 mm
Diameter (Metric) Inside 100 mm
Product Content Correction
Product Title
Cytiva Whatman™ WT 100 Witt's Flask >