Learn More
Tocris Bioscience™ WAY 100635 maleate
GREENER_CHOICE
Product Code. 18085952
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18085952
Brand: Tocris Bioscience™ 4380/10
Potent 5-HT1A antagonist; also D4 agonist
Potent, silent antagonist of 5-HT1A receptors (IC50 = 2.2 nM; Ki = 0.84 nM for rat 5-HT1A receptors). Displays 100-fold selectivity for 5-HT1A over other 5-HT subtypes. Also exhibits agonist activity at dopamine D4 receptors.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 634908-75-1
Molecular Formula C29H40N4O6
Specifications
Quantity 10 mg
Formula Weight 538.64
Purity >99%
Molecular Weight (g/mol) 540.661
Chemical Name or Material WAY 100635 maleate