Learn More
Tocris Bioscience™ CPYPP
Description
CPYPP is a DOCK2-Rac1 interaction inhibitor; suppresses DOCK2-induced Rac1 activation. Also inhibits DOCK1 and DOCK5. Exhibits selectivity over DOCK9. Reduces chemotactic responses in vitro and in vivo, and activates T cells in vitro. Also attenuates HER2-mediated breast cancer cell migration in vitro.
Specifications
Specifications
| Chemical Name or Material | Rho GTPase Inhibitors |
| CAS | 310460-39-0 |
| Quantity | 10 mg |
| Target | Small G Protein Inhibitors |
| Molecular Formula | C18H13ClN2O2 |
| Formula Weight | 324.76 |
| Purity | >98% |