Learn More
Tocris Bioscience™ VH 101, thiol
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
VH 101, thiol is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand with terminal thiol ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-1-[(2R)-2-[[(1-Fluorocyclopropyl)carbonyl]amino]-3-methyl-3-sulfanyl-butanoyl]-4-hydroxy-N-[[4-(4-methylthiazol-5-yl)phenyl]methyl]pyrrolidine-2-carboxamide |
| CAS | 2358775-81-0 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C25H31FN4O4S2 |
| Purity | 0.95 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur