missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ VER 246608
GREENER_CHOICE

Product Code. 18759926
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759926

Brand: Tocris Bioscience™ 6604/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3/4 inhibitor

VER 246608 is a potent and ATP-competitive inhibitor of pyruvate dehydrogenase kinase (PDK; IC50s = 35, 40, 84, and 91 nM for PDK-1, PDK-3, PDK-2, and PDK-4 respectively). It is selective for PDKs over a panel of 97 kinases and HSP90. VER 246608 increases pyruvate dehydrogenase complex activity, oxygen consumption and reduces glycolytic activity; it reduces the production of L-lactate in PC3 cells in D-glucose and L-glutamine depleted media. In PC3 cells, VER 246608 induces cell cycle arrest at the G1 phase under glucose or serum-depleted conditions characteristic of the tumor microenvironment.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-(2-Chloro-5-methyl-4-pyrimidinyl)phenyl]-N-[[4-[[(2,2-difluoroacetyl)amino]methyl]phenyl]methyl]-2,4-dihydroxybenzamide
CAS 1684386-71-7
Quantity 50 mg
Target Other Kinase Inhibitors
Molecular Formula C28H23ClF2N4O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.