missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM3 Rat anti-Mouse, Clone: 295502, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2224-SP

This item is not returnable.
View return policy
Description
TREM3 Monoclonal specifically detects TREM3 in Mouse samples. It is validated for Western Blot.Specifications
TREM3 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_067382 | |
Trem3 | |
Mouse myeloma cell line NS0-derived recombinant mouse TREM-3 Met1-Pro135 (predicted) Accession # NP_067382 | |
25 μg | |
Primary | |
Detects mouse TREM-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) TREM-1 or rmTREM-2b is observed. | |
Mouse | |
Purified |
Western Blot | |
295502 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BB134760, TREM3 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54210 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
TREM3 Rat anti-Mouse, Clone: 295502, R&D Systems™
Spot an opportunity for improvement?Share a Content Correction