Learn More
Tocris Bioscience™ Tocriscreen Epigenetics 3.0
Product Code. 18792344
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
Product Code. 18792344
Brand: Tocris Bioscience™ 7578/1 SET
A library of 160 epigenetic compounds (100 μL, 10 mM DMSO solutions) for high throughput screening, high content screening, target validation and assay development
Tocriscreen Epigenetics Library is a library of 160 epigenetic compounds (100 μL, 10 mM solutions). Contains compounds covering >40 epigenetic targets including epigenetic readers, writers, erasers and transcriptional modulators. Includes the latest chemical tools to target p300/CBP, PCAF, BRPF1 and SMYD2.This library replaces the Tocriscreen Epigenetics Library (Cat. No. 6801).If this library does not suit your needs, please submit your requirements through our Tocriscreen PRO custom compound library service .
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Set
Target Screening Libraries