Learn More
STEMCELL Technologies Tivantinib
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- Tivantinib is a staurosporine derivative which binds to inactive, dephosphorylated MET receptor tyrosine kinase, in a manner that is not competitive with ATP (Munshi et al.; Eathiraj et al.) It is selective for MET (Ki ≅ 355 nM) in a screen of 230 kinases (Munshi et al.) It also shows cytotoxic activity which is distinct from its inhibition of MET (Basilico et al.; Katayama et al.) It has also been shown to bind directly to the colchicine binding pocket of tubulin, thereby reducing tubulin polymerization (Aoyama et al.)
Specifications
Specifications
| CAS | 905854-02-6 |
| Synonym | ARQ 197 |
| Quantity | 10 mg |
| Molecular Formula | C23H19N3O2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.