Learn More
Thermo Scientific™ ThermoChill II Low Range Recirculating Chiller
Low Range chillers support applications requiring temperatures of -10 °C to 30 °C
Brand:  Thermo Scientific™ 197221120000
51825.59 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15321657
  • 52084.72 NOK / Each
Only null left
Estimated Shipment: 11-12-2025
Product Title
Thermo Scientific™ ThermoChill II Low Range Recirculating Chiller > Low range; 230 V/ 50Hz; PD-1 RS 232