Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC4-acid
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Thalidomide 4'-ether-alkylC4-acid is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkylC4 linker with a terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 5-[[2-(2,6-Dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]pentanoic acid |
| CAS | 2169266-67-3 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C18H18N2O7 |
| Purity | 0.95 |