missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies TG101348

Product Code. p-7217484
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17110232

Brand: STEMCELL Technologies 73472

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

JAK/STAT pathway inhibitor; Inhibits JAK2

  • TG101348 is an inhibitor of Janus kinase 2 (JAK2) with an IC50 of 6 nM
  • It also inhibits related kinases fms-related tyrosine kinase 3 (FLT3), RET, and JAK3 with less potent activity, having IC50 values of 25, 17 and 169 nM, respectively (Pardanani et al.) It is proposed to bind to the ATP binding pocket of its kinase targets (Zhou et al.)
  • Inhibits growth of Ba/F3 cells expressing JAK2 V617F or MPL W515L mutations (Pardanani et al.)
  • Reduces tumor cell burden and increases survival in mouse models of JAK2 V617F-induced hematopoietic and myeloproliferative disease (Pardanani et al.; Wernig et al.)
  • Sensitizes erlotinib-resistant non-small cell lung cancer cells to erlotinib treatment in vitro and in a mouse xenograft model (Zhang et al.)
  • Displaces BRD4 from chromatin and suppresses c-MYC expression in multiple myeloma cells in vitro (Ciceri et al.)

Specifications

CAS 936091-26-8
Synonym Fedratinib; SAR302503
Quantity 1 mg
Molecular Formula C27H36N6O3S
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.